siRNA Details
| Gene Symbol: Slco2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033314.3 | NP_201571.2 | 6 | 935 | ||
| GenBank mRNA | AF323958.1 | - | 6 | 804 | ||
| AK087350.1 | - | 6 | 941 | |||
| AK166724.1 | - | 6 | 936 | |||
| AK169706.1 | - | 6 | 938 | |||
| BC035200.1 | - | 925 | 3158233 | |||

Slco2a1
Genomic Map