siRNA Details
| Gene Symbol: Derl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291146.1 | NP_001278075.1 | 5 | 732 | ||
| NM_001291147.1 | NP_001278076.1 | 5 | 476 | |||
| NM_001291148.1 | NP_001278077.1 | 5 | 438 | |||
| NM_033562.4 | NP_291040.1 | 5 | 441 | |||
| GenBank mRNA | AF208064.1 | - | 5 | 380 | ||
| AK048627.1 | - | 5 | 388 | |||
| AK077659.1 | - | 5 | 392 | |||
| AK082564.1 | - | 5 | 401 | |||
| AK083632.1 | - | 5 | 380 | |||
| AK083725.1 | - | 2 | 3144 | |||
| AK090105.1 | - | 5 | 450 | |||
| AK151934.1 | - | 5 | 732 | |||
| BC005682.1 | - | 5 | 380 | 3154448 | ||
| BC037503.1 | - | 1 | 23 | |||
| BE627680.1 | - | 5 | 409 | |||

Derl2
Genomic Map