siRNA Details
| Gene Symbol: Pcdhgc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033583.3 | NP_291061.1 | 1 | 752 | ||
| GenBank mRNA | AF464160.1 | - | 1 | 648 | ||
| AF464163.1 | - | 1 | 752 | |||
| AY013813.1 | - | 1 | 623 | |||
| BC138677.1 | - | 1 | 696 | 8861698 | ||
| BC138678.1 | - | 1 | 810 | 8861699 | ||

Pcdhgc5
Genomic Map