siRNA Details
| Gene Symbol: Mbd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033072.2 | NP_149063.2 | 13 | 3595 | ||
| GenBank mRNA | AK018262.1 | - | 4 | 736 | ||
| AK047331.1 | - | 3 | 620 | |||
| AK158797.1 | - | 13 | 3713 | |||
| BC019433.1 | - | 5 | 1096 | |||
| BC049833.1 | - | 5 | 1079 | |||
| BC055445.1 | - | 5 | 1052 | |||
| BC059015.1 | - | 14 | 3378 | |||
| BC083328.1 | - | 4 | 732 | |||
| BC096027.1 | - | 10 | 3337 | |||

Mbd6
Genomic Map