siRNA Details
| Gene Symbol: Zfp423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033327.2 | NP_201584.2 | 6 | 3975 | ||
| GenBank mRNA | AF188609.1 | - | 4 | 3344 | ||
| AK122365.1 | - | 6 | 3941 | |||
| AK133497.1 | - | 4 | 3403 | |||
| AY147407.1 | - | 5 | 3580 | |||
| AY256893.1 | - | 6 | 3874 | |||
| BC059234.1 | - | 4 | 3786 | |||
| BC079586.1 | - | 6 | 3661 | |||
| BC139028.1 | - | 6 | 3721 | 8862050 | ||
| BC139030.1 | - | 6 | 3721 | 8862052 | ||
| BU519630.1 | - | 3 | 198 | |||

Zfp423
Genomic Map