siRNA Details
| Gene Symbol: Derl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291146.1 | NP_001278075.1 | 7 | 3110 | ||
| NM_001291147.1 | NP_001278076.1 | 7 | 2854 | |||
| NM_001291148.1 | NP_001278077.1 | 7 | 2816 | |||
| NM_033562.4 | NP_291040.1 | 7 | 2819 | |||
| GenBank mRNA | AK082564.1 | - | 7 | 2779 | ||
| AK090105.1 | - | 7 | 2827 | |||

Derl2
Genomic Map