siRNA Details
| Gene Symbol: Myb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198914.1 | NP_001185843.1 | 4 | 551 | ||
| NM_010848.3 | NP_034978.3 | 4 | 551 | |||
| GenBank mRNA | AK088079.1 | - | 4 | 472 | ||
| AK088598.1 | - | 4 | 473 | |||
| AK132250.1 | - | 4 | 416 | |||
| AK137965.1 | - | 4 | 417 | |||
| BC011513.1 | - | 4 | 459 | 3672769 | ||
| M12848.1 | - | 4 | 549 | |||
| X02774.1 | - | 4 | 321 | |||

Myb
Genomic Map