siRNA Details
| Gene Symbol: Adam33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163529.1 | NP_001157001.1 | 3 | 536 | ||
| NM_033615.2 | NP_291093.2 | 3 | 536 | |||
| GenBank mRNA | AB059632.1 | - | 3 | 536 | ||
| AB059633.1 | - | 3 | 536 | |||
| AF472524.1 | - | 3 | 268 | |||
| AK053825.1 | - | 4 | 340 | |||
| AK086734.1 | - | 3 | 237 | |||
| AY054984.1 | - | 3 | 236 | |||
| AY382193.1 | - | 3 | 267 | |||
| AY382194.1 | - | 3 | 267 | |||
| BC138139.1 | - | 3 | 259 | 8861158 | ||

Adam33
Genomic Map