siRNA Details
Gene Symbol: Klhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053105.2 | NP_444335.2 | 1 | 1084 | ||
GenBank mRNA | AF252281.1 | - | 1 | 1013 | ||
AK031932.1 | - | 1 | 896 | |||
AK220397.1 | - | 1 | 388 | |||
BC094584.1 | - | 1 | 1084 | 30547502 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
