siRNA Details
Gene Symbol: Rnase2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053113.2 | NP_444343.2 | 2 | 228 | ||
GenBank mRNA | BC020070.1 | - | 2 | 227 | 3982432 | |
BC027557.1 | - | 2 | 228 | 1445853 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rnase2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053113.2 | NP_444343.2 | 2 | 228 | ||
GenBank mRNA | BC020070.1 | - | 2 | 227 | 3982432 | |
BC027557.1 | - | 2 | 228 | 1445853 |