siRNA Details
| Gene Symbol: Gprc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205396.1 | NP_001192325.1 | 2 | 618 | ||
| NM_053118.1 | NP_444348.1 | 2 | 618 | |||
| GenBank mRNA | AB025221.1 | - | 1 | 464 | ||
| AB025222.1 | - | 1 | 464 | |||
| AB099816.1 | - | 1 | 509 | |||
| AF218809.1 | - | 2 | 618 | |||
| AK137739.1 | - | 2 | 581 | |||
| BC119192.1 | - | 2 | 578 | 8733941 | ||
| BC119194.1 | - | 2 | 578 | 8733943 | ||
More Information
Set Membership: |
|

Gprc5d
Genomic Map