siRNA Details
| Gene Symbol: Dxo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163770.1 | NP_001157242.1 | 3 | 609 | ||
| NM_033613.2 | NP_291091.2 | 3 | 634 | |||
| GenBank mRNA | BC004713.1 | - | 3 | 570 | 3499698 | |
| BX522724.1 | - | 3 | 523 | |||

Dxo
Genomic Map