siRNA Details
| Gene Symbol: Agpat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053014.3 | NP_443747.2 | 6 | 721 | ||
| GenBank mRNA | AB377215.1 | - | 4 | 537 | ||
| AK008965.1 | - | 6 | 877 | |||
| AK015906.1 | - | 5 | 649 | |||
| AK030607.1 | - | 4 | 513 | |||
| AK075715.1 | - | 6 | 721 | |||
| AK076414.1 | - | 5 | 690 | |||
| AK159110.1 | - | 5 | 961 | |||
| AY167588.1 | - | 4 | 559 | |||
| BC033444.1 | - | 4 | 399 | |||
| BC052382.1 | - | 5 | 612 | 5703571 | ||
| BC058519.1 | - | 5 | 611 | 5703571 | ||

Agpat3
Genomic Map