siRNA Details
| Gene Symbol: Adarb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289530.1 | NP_001276459.1 | 10 | 3214 | ||
| NM_052977.5 | NP_443209.2 | 10 | 3219 | |||
| GenBank mRNA | AK033916.1 | - | 10 | 3158 | ||
| AK045538.1 | - | 10 | 3159 | |||
| AK046083.1 | - | 10 | 3158 | |||
| AK134964.1 | - | 10 | 3221 | |||
| AK139079.1 | - | 10 | 3177 | |||
| AK139273.1 | - | 10 | 3177 | |||
| BC052426.1 | - | 10 | 3144 | 6835942 | ||

Adarb2
Genomic Map