siRNA Details
| Gene Symbol: Epgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053087.2 | NP_444317.1 | 5 | 906 | ||
| GenBank mRNA | AJ291391.1 | - | 5 | 906 | ||
| AK010136.1 | - | 4 | 854 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epgn| Gene Symbol: Epgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053087.2 | NP_444317.1 | 5 | 906 | ||
| GenBank mRNA | AJ291391.1 | - | 5 | 906 | ||
| AK010136.1 | - | 4 | 854 | |||