siRNA Details
| Gene Symbol: Trim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053100.2 | NP_444330.2 | 6 | 2951 | ||
| GenBank mRNA | AF281047.1 | - | 6 | 2300 | ||
| AK052763.1 | - | 6 | 1688 | |||
| AK079847.1 | - | 4 | 1394 | |||
| AK155303.1 | - | 6 | 2896 | |||
| BC060094.1 | - | 6 | 2182 | |||
| BC082298.1 | - | 6 | 2826 | |||

Trim8
Genomic Map