siRNA Details
| Gene Symbol: Pcdhb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053127.2 | NP_444357.1 | 1 | 1089 | ||
| GenBank mRNA | AY013781.1 | - | 1 | 953 | ||
| BC141154.1 | - | 1 | 1045 | 9055766 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhb2| Gene Symbol: Pcdhb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053127.2 | NP_444357.1 | 1 | 1089 | ||
| GenBank mRNA | AY013781.1 | - | 1 | 953 | ||
| BC141154.1 | - | 1 | 1045 | 9055766 | ||