siRNA Details
| Gene Symbol: Klra21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053151.1 | NP_444381.1 | 513 | |||
| GenBank mRNA | AF288380.1 | - | 513 | |||
| AY971806.1 | - | 407 | ||||
| BC116399.1 | - | 481 | 40101978 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klra21| Gene Symbol: Klra21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053151.1 | NP_444381.1 | 513 | |||
| GenBank mRNA | AF288380.1 | - | 513 | |||
| AY971806.1 | - | 407 | ||||
| BC116399.1 | - | 481 | 40101978 | |||