siRNA Details
| Gene Symbol: Ugt2b37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053215.3 | NP_444445.2 | 1 | 302 | ||
| GenBank mRNA | AK002736.1 | - | 1 | 319 | ||
| BC019487.1 | - | 1 | 302 | 4237718 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ugt2b37| Gene Symbol: Ugt2b37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053215.3 | NP_444445.2 | 1 | 302 | ||
| GenBank mRNA | AK002736.1 | - | 1 | 319 | ||
| BC019487.1 | - | 1 | 302 | 4237718 | ||