siRNA Details
Gene Symbol: Vmn1r42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053221.2 | NP_444451.2 | 1 | 172 | ||
GenBank mRNA | BC125372.1 | - | 1 | 172 | 40129887 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn1r42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053221.2 | NP_444451.2 | 1 | 172 | ||
GenBank mRNA | BC125372.1 | - | 1 | 172 | 40129887 |