siRNA Details
| Gene Symbol: Vmn1r52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053222.1 | NP_444452.1 | 1 | 830 | ||
| Gene Symbol: V1ra8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC138392.1 | - | 1 | 855 | 8861412 | |
| BC138394.1 | - | 1 | 855 | 8861414 | ||

Vmn1r52
Genomic Map