siRNA Details
Gene Symbol: Vmn1r53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053226.2 | NP_444456.2 | 1 | 681 | ||
GenBank mRNA | BC125605.1 | - | 1 | 681 | 40130120 | |
BC132002.1 | - | 1 | 681 | 40130279 |
