siRNA Details
| Gene Symbol: Vmn1r29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053232.1 | NP_444462.1 | 1 | 482 | ||
| GenBank mRNA | BC150722.1 | - | 1 | 511 | 9087633 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r29| Gene Symbol: Vmn1r29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053232.1 | NP_444462.1 | 1 | 482 | ||
| GenBank mRNA | BC150722.1 | - | 1 | 511 | 9087633 | |