siRNA Details
| Gene Symbol: Foxp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286607.1 | NP_001273536.1 | 3 | 507 | ||
| NM_053242.4 | NP_444472.2 | 5 | 844 | |||
| NM_212435.1 | NP_997600.1 | 3 | 507 | |||
| GenBank mRNA | AF339106.1 | - | 2 | 185 | ||
| AK051668.1 | - | 5 | 806 | |||
| AK080549.1 | - | 5 | 715 | |||
| AK082361.1 | - | 5 | 516 | |||
| AY079003.1 | - | 5 | 473 | |||
| BC058960.1 | - | 3 | 504 | 6811877 | ||
| BC062926.1 | - | 3 | 507 | 6851153 | ||

Foxp2
Genomic Map