siRNA Details
| Gene Symbol: Dok4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053246.3 | NP_444476.1 | 3, 4 | 507 | ||
| GenBank mRNA | AF418207.1 | - | 2, 3 | 391 | ||
| AK032352.1 | - | 3, 4 | 497 | |||
| AK043670.1 | - | 2, 3 | 419 | |||
| AK077612.1 | - | 3, 4 | 498 | |||
| AK139363.1 | - | 2, 3 | 395 | |||
| AK142621.1 | - | 2, 3 | 414 | |||
| AK156485.1 | - | 3, 4 | 507 | |||
| BC004705.1 | - | 3, 4 | 468 | 3499419 | ||

Dok4
Genomic Map