siRNA Details
| Gene Symbol: Crip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053250.2 | NP_444480.1 | 4, 5 | 346 | ||
| NM_181664.2 | NP_858050.1 | 4, 5 | 346 | |||
| GenBank mRNA | AF367970.1 | - | 4, 5 | 346 | ||
| AF367971.1 | - | 4, 5 | 346 | |||
| AK080272.1 | - | 4, 5 | 346 | |||
| AY555745.1 | - | 4, 5 | 337 | |||
| BC116360.1 | - | 4, 5 | 346 | 40099973 | ||

Crip3
Genomic Map