siRNA Details
| Gene Symbol: Pcdhb13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053138.2 | NP_444368.1 | 1 | 305 | ||
| GenBank mRNA | AK082435.1 | - | 1 | 307 | ||
| AY013774.1 | - | 1 | 251 | |||
| BC138123.1 | - | 1 | 322 | 8861142 | ||
| BC138124.1 | - | 1 | 322 | 8861143 | ||

Pcdhb13
Genomic Map