siRNA Details
| Gene Symbol: Clmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040682.1 | NP_001035772.1 | 2 | 240 | ||
| NM_053155.2 | NP_444385.2 | 2 | 240 | |||
| GenBank mRNA | AB047978.1 | - | 2 | 233 | ||
| AB059643.1 | - | 2 | 233 | |||
| AB059644.1 | - | 2 | 233 | |||
| AB059645.1 | - | 2 | 233 | |||
| AB059646.1 | - | 2 | 233 | |||
| AB059647.1 | - | 2 | 233 | |||
| AB059648.1 | - | 2 | 233 | |||
| AK147777.1 | - | 2 | 137 | |||
| BC019176.1 | - | 2 | 171 | |||
| BC096470.1 | - | 2 | 129 | |||
| BC141280.1 | - | 2 | 173 | 9055892 | ||

Clmn
Genomic Map