siRNA Details
| Gene Symbol: Mrpl43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053164.3 | NP_444394.1 | 3 | 887 | ||
| GenBank mRNA | AA253916.1 | - | 1 | 149 | ||
| AK076204.1 | - | 3 | 846 | |||
| BC065812.1 | - | 3 | 834 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl43| Gene Symbol: Mrpl43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053164.3 | NP_444394.1 | 3 | 887 | ||
| GenBank mRNA | AA253916.1 | - | 1 | 149 | ||
| AK076204.1 | - | 3 | 846 | |||
| BC065812.1 | - | 3 | 834 | |||