siRNA Details
| Gene Symbol: Ric8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053194.4 | NP_444424.1 | 3, 4 | 792 | ||
| GenBank mRNA | AF288813.1 | - | 3, 4 | 791 | ||
| AK031466.1 | - | 3, 4 | 739 | |||
| AK080192.1 | - | 3, 4 | 735 | |||
| AK155282.1 | - | 3, 4 | 767 | |||
| AK165536.1 | - | 3, 4 | 769 | |||
| AK167746.1 | - | 3, 4 | 795 | |||
| AK169367.1 | - | 3, 4 | 756 | |||
| BC022917.1 | - | 3, 4 | 769 | 4460521 | ||

Ric8
Genomic Map