siRNA Details
| Gene Symbol: Magee1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053201.3 | NP_444431.3 | 1 | 1459 | ||
| GenBank mRNA | AF319978.1 | - | 1 | 662 | ||
| AK030032.1 | - | 1 | 1453 | |||
| AK051625.1 | - | 1 | 1433 | |||
| AK081378.1 | - | 1 | 1435 | |||
| BC059039.1 | - | 1 | 1459 | 6812964 | ||

Magee1
Genomic Map