siRNA Details
| Gene Symbol: Vmn1r40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053228.1 | NP_444458.1 | 1 | 583 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r40| Gene Symbol: Vmn1r40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053228.1 | NP_444458.1 | 1 | 583 | ||