siRNA Details
| Gene Symbol: Dok4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053246.3 | NP_444476.1 | 5 | 630 | ||
| GenBank mRNA | AF418207.1 | - | 4 | 514 | ||
| AK032352.1 | - | 5 | 620 | |||
| AK043670.1 | - | 4 | 542 | |||
| AK077612.1 | - | 5 | 622 | |||
| AK139363.1 | - | 4 | 518 | |||
| AK142621.1 | - | 4 | 537 | |||
| AK156485.1 | - | 5 | 630 | |||
| AK192791.1 | - | 2 | 114 | |||
| BC004705.1 | - | 5 | 591 | 3499419 | ||

Dok4
Genomic Map