siRNA Details
| Gene Symbol: Ehbp1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114597.1 | NP_001108069.1 | 9 | 1292 | ||
| NM_053252.3 | NP_444482.2 | 9 | 1292 | |||
| GenBank mRNA | AF305087.1 | - | 9 | 1245 | ||
| AF305088.1 | - | 9 | 1245 | |||
| AK087817.1 | - | 5 | 534 | |||
| AK169974.1 | - | 9 | 1258 | |||
| BC004660.1 | - | 9 | 1241 | 3497913 | ||

Ehbp1l1
Genomic Map