siRNA Details
| Gene Symbol: Pcdhb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053135.2 | NP_444365.1 | 1 | 1134 | ||
| GenBank mRNA | AY013771.1 | - | 1 | 935 | ||
| BC112378.1 | - | 1 | 930 | |||
| BC112397.1 | - | 1 | 934 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhb10| Gene Symbol: Pcdhb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053135.2 | NP_444365.1 | 1 | 1134 | ||
| GenBank mRNA | AY013771.1 | - | 1 | 935 | ||
| BC112378.1 | - | 1 | 930 | |||
| BC112397.1 | - | 1 | 934 | |||