siRNA Details
| Gene Symbol: Hemgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053149.2 | NP_444379.1 | 3 | 403 | ||
| GenBank mRNA | AF269248.1 | - | 3 | 403 | ||
| AK076587.1 | - | 4 | 388 | |||
| AK077692.1 | - | 3 | 246 | |||
| AK156536.1 | - | 3 | 331 | |||
| AK171890.1 | - | 3 | 322 | |||
| AK171906.1 | - | 3 | 452 | |||
| AK172421.1 | - | 3 | 322 | |||
| AY699986.1 | - | 3 | 245 | |||
| BC050798.1 | - | 4 | 382 | 6743819 | ||

Hemgn
Genomic Map