siRNA Details
| Gene Symbol: Prss28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053259.2 | NP_444489.2 | 3 | 304 | ||
| GenBank mRNA | AF184895.1 | - | 2 | 143 | ||
| AK144498.1 | - | 3 | 305 | |||
| BC119386.1 | - | 2 | 171 | 8734135 | ||
| BC119388.1 | - | 2 | 171 | 8734137 | ||

Prss28
Genomic Map