siRNA Details
| Gene Symbol: Selm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053267.2 | NP_444497.1 | 2 | 390 | ||
| GenBank mRNA | AK018791.1 | - | 2 | 226 | ||
| AK035312.1 | - | 2 | 180 | |||
| AK046034.1 | - | 2 | 182 | |||
| AK079358.1 | - | 2 | 208 | |||
| AK079576.1 | - | 2 | 409 | |||
| AY043488.1 | - | 2 | 206 | |||
| BC019742.1 | - | 2 | 82 | |||
| BC147332.1 | - | 2 | 300 | 9056406 | ||
| BC147333.1 | - | 2 | 302 | 9056396 | ||
| BI731890.1 | - | 2 | 388 | |||
| CK787580.1 | - | 2 | 382 | |||

Selm
Genomic Map