siRNA Details
| Gene Symbol: Tulp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054040.3 | NP_473381.1 | 1 | 858 | ||
| GenBank mRNA | AF219945.1 | - | 1 | 262 | ||
| AK173165.1 | - | 1 | 858 | |||
| BC138024.1 | - | 1 | 193 | 8861043 | ||
| BC138025.1 | - | 1 | 193 | 8861044 | ||

Tulp4
Genomic Map