siRNA Details
| Gene Symbol: Cd248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054042.2 | NP_473383.1 | 1 | 927 | ||
| GenBank mRNA | AF378758.1 | - | 1 | 885 | ||
| AF388572.1 | - | 1 | 882 | |||
| AK144655.1 | - | 1 | 897 | |||
| BC046318.1 | - | 1 | 927 | 6314798 | ||

Cd248
Genomic Map