siRNA Details
| Gene Symbol: Def8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253783.1 | NP_001240712.1 | 4 | 297 | ||
| NM_001253784.1 | NP_001240713.1 | 3 | 275 | |||
| NM_001281803.1 | NP_001268732.1 | 2 | 279 | |||
| NM_054046.5 | NP_473387.1 | 2 | 188 | |||
| GenBank mRNA | AK028726.1 | - | 3 | 247 | ||
| AK034028.1 | - | 154 | ||||
| AK046271.1 | - | 2 | 149 | |||
| AK050405.1 | - | 3 | 246 | |||
| AK081295.1 | - | 2 | 178 | |||
| AK086174.1 | - | 2 | 149 | |||
| AK087778.1 | - | 2 | 159 | |||
| AK129019.1 | - | 2 | 230 | |||
| AK133979.1 | - | 2 | 162 | |||
| AK150334.1 | - | 2 | 124 | |||
| AK150994.1 | - | 2 | 124 | |||
| AK164147.1 | - | 2 | 160 | |||
| BC003306.1 | - | 4 | 248 | 3154712 | ||
| BY069015.1 | - | 3 | 171 | |||
| BY273027.1 | - | 2 | 238 | |||
| BY285979.1 | - | 2 | 279 | |||
| BY713285.1 | - | 2 | 189 | |||
| CX562217.1 | - | 2 | 241 | |||
| X96707.1 | - | 2 | 180 | |||

Def8
Genomic Map