siRNA Details
| Gene Symbol: Prosc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039077.2 | NP_001034166.1 | 3 | 281 | ||
| NM_054057.4 | NP_473398.1 | 3 | 281 | |||
| GenBank mRNA | AK032308.1 | - | 3 | 231 | ||
| AK049905.1 | - | 3 | 240 | |||
| AK134549.1 | - | 3 | 214 | |||
| AK147670.1 | - | 3 | 263 | |||
| AK160172.1 | - | 3 | 235 | |||
| AK203439.1 | - | 3 | 170 | |||
| BC061045.1 | - | 3 | 275 | 6534037 | ||
| BY065041.1 | - | 3 | 281 | |||

Prosc
Genomic Map