siRNA Details
| Gene Symbol: Pip4k2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054097.3 | NP_473438.1 | 2 | 368 | ||
| GenBank mRNA | AB054591.1 | - | 2 | 311 | ||
| AK140079.1 | - | 2 | 332 | |||
| AK148299.1 | - | 2 | 328 | |||
| AK182167.1 | - | 1 | 61 | |||
| BC021383.1 | - | 2 | 301 | 5042933 | ||

Pip4k2c
Genomic Map