siRNA Details
| Gene Symbol: 1110038F14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054099.2 | NP_473440.1 | 4 | 832 | ||
| GenBank mRNA | AK004154.1 | - | 4 | 786 | ||
| AK141496.1 | - | 4 | 624 | |||
| AK153059.1 | - | 5 | 786 | |||
| AK160695.1 | - | 4 | 855 | |||
| BC038324.1 | - | 4 | 832 | 1511597 | ||
| BC091734.1 | - | 4 | 597 | 3663572 | ||

1110038F14Rik
Genomic Map