siRNA Details
| Gene Symbol: C1QB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000491.3 | NP_000482.3 | 2 | 309 | ||
| XM_011542059.2 | XP_011540361.1 | 3 | 460 | |||
| GenBank mRNA | BC008983.1 | - | 2 | 251 | 4212848 | |
| BQ711602.1 | - | 3 | 471 | |||
| X03084.1 | - | 1 | 103 | |||

C1QB
Genomic Map