siRNA Details
| Gene Symbol: Zfp358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080461.2 | NP_536709.2 | 2 | 1049 | ||
| GenBank mRNA | AF397208.1 | - | 2 | 1014 | ||
| AK155446.1 | - | 2 | 1051 | |||
| BC035489.1 | - | 1 | 605 | |||
| BC040067.1 | - | 1 | 723 | 4008695 | ||
| BC055855.1 | - | 1 | 1250 | |||

Zfp358
Genomic Map