siRNA Details
| Gene Symbol: Slc13a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054055.2 | NP_473396.1 | 2 | 159 | ||
| GenBank mRNA | AF306491.1 | - | 2 | 158 | ||
| AK085326.1 | - | 2 | 157 | |||
| AK086775.1 | - | 2 | 161 | |||
| AK087518.1 | - | 2 | 157 | |||
| AK128982.1 | - | 2 | 156 | |||
| AK138344.1 | - | 2 | 157 | |||
| AK157881.1 | - | 2 | 128 | |||
| AK165355.1 | - | 2 | 157 | |||
| BC026803.1 | - | 2 | 145 | 4238769 | ||
| BC028882.1 | - | 2 | 143 | |||

Slc13a3
Genomic Map