siRNA Details
| Gene Symbol: Calcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054084.2 | NP_473425.2 | 3 | 263 | ||
| GenBank mRNA | AK136518.1 | - | 3 | 257 | ||
| BC111865.1 | - | 3 | 263 | 40049329 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Calcb| Gene Symbol: Calcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054084.2 | NP_473425.2 | 3 | 263 | ||
| GenBank mRNA | AK136518.1 | - | 3 | 257 | ||
| BC111865.1 | - | 3 | 263 | 40049329 | ||