siRNA Details
| Gene Symbol: Prss29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053260.3 | NP_444490.2 | 5 | 718 | ||
| GenBank mRNA | AF305425.2 | - | 4 | 554 | ||
| AF442819.1 | - | 4 | 554 | |||
| AK144368.1 | - | 5 | 719 | |||
| BC104122.1 | - | 4 | 554 | 40044207 | ||
| BC104123.1 | - | 4 | 554 | |||

Prss29
Genomic Map