siRNA Details
| Gene Symbol: Selm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053267.2 | NP_444497.1 | 3, 4 | 437 | ||
| GenBank mRNA | AK018791.1 | - | 3, 4 | 273 | ||
| AK035312.1 | - | 3, 4 | 227 | |||
| AK046034.1 | - | 3, 4 | 229 | |||
| AK079358.1 | - | 3, 4 | 255 | |||
| AY043488.1 | - | 3, 4 | 253 | |||
| BC019742.1 | - | 3, 4 | 129 | |||
| BC147332.1 | - | 3, 4 | 347 | 9056406 | ||
| BC147333.1 | - | 3, 4 | 349 | 9056396 | ||
| CK787580.1 | - | 3, 4 | 429 | |||

Selm
Genomic Map